Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005544800.2 | XP_005544857.1 | 3 - 4 | 677 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |