Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7H14orf28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015453738.1 | XP_015309224.1 | 2 - 3 | 546 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |