Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005567456.2 | XP_005567513.1 | 3 - 4 | 448 | 74 | |
XM_005567457.2 | XP_005567514.1 | 3 - 4 | 446 | 74 | ||
XM_005567458.2 | XP_005567515.1 | 3 - 4 | 429 | 74 | ||
XM_015457260.1 | XP_015312746.1 | 2 - 3 | 482 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |