Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUXA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005590495.1 | XP_005590552.1 | 2 - 3 | 175 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |