Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005585050.2 | XP_005585107.1 | 2 - 3 | 853 | 65 | |
XM_015438871.1 | XP_015294357.1 | 1 - 2 | 1158 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Inventoried Probe spans exons |