Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005574706.2 | XP_005574763.1 | 1 - 2 | 931 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |