Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005580103.2 | XP_005580160.1 | 4 - 5 | 1361 | 104 | |
XM_015435917.1 | XP_015291403.1 | 4 - 5 | 1491 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |