Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARVELD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005592442.1 | XP_005592499.1 | 1 - 2 | 489 | 77 | |
XM_005592443.2 | XP_005592500.1 | 1 - 2 | 489 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |