Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005565098.2 | XP_005565155.1 | 4 - 5 | 690 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |