Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C13H2orf78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005575584.2 | XP_005575641.1 | 2 - 3 | 990 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |