Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSGN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005576495.2 | XP_005576552.1 | 1 | 745 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |