Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005572402.2 | XP_005572459.1 | 2 - 3 | 301 | 82 | |
XM_005572403.2 | XP_005572460.1 | 2 - 3 | 304 | 82 | ||
GenBank mRNA | AB056397.1 | - | 2 - 3 | 237 | 82 | |
AB168826.1 | - | 2 - 3 | 238 | 82 | ||
AB172295.1 | - | 2 - 3 | 238 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |