Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005590115.2 | XP_005590172.1 | 5 - 6 | 1475 | 66 | |
XM_005590116.2 | XP_005590173.1 | 4 - 5 | 1094 | 66 | ||
XM_015441931.1 | XP_015297417.1 | 4 - 5 | 1094 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |