Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005543841.2 | XP_005543898.1 | 1 - 2 | 342 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |