Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005587527.2 | XP_005587584.1 | 1 - 2 | 165 | 62 | |
XM_005587528.2 | XP_005587585.1 | 1 - 2 | 165 | 62 | ||
XM_005587529.2 | XP_005587586.1 | 1 - 2 | 165 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |