Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005590083.2 | XP_005590140.1 | 3 - 4 | 1273 | 85 | |
XM_005590084.2 | XP_005590141.1 | 3 - 4 | 1286 | 85 | ||
XM_005590086.2 | XP_005590143.1 | 3 - 4 | 1274 | 85 | ||
XM_005590089.2 | XP_005590146.1 | 3 - 4 | 1312 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |