Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102130068 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015442186.1 | XP_015297672.1 | 15 - 16 | 2052 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |