Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5H4orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005556434.2 | XP_005556491.1 | 2 - 3 | 494 | 108 | |
XM_005556436.2 | XP_005556493.1 | 2 - 3 | 497 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |