Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTIF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005585529.2 | XP_005585586.1 | 3 - 4 | 779 | 97 | |
XM_005585531.2 | XP_005585588.1 | 3 - 4 | 857 | 97 | ||
XM_015439097.1 | XP_015294583.1 | 4 - 5 | 849 | 97 | ||
XM_015439098.1 | XP_015294584.1 | 4 - 5 | 925 | 97 | ||
XM_015439099.1 | XP_015294585.1 | 4 - 5 | 1271 | 97 | ||
XM_015439100.1 | XP_015294586.1 | 4 - 5 | 816 | 97 | ||
XM_015439101.1 | XP_015294587.1 | 4 - 5 | 975 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |