Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEM1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005587586.2 | XP_005587643.1 | 1 | 2096 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |