Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDR9C7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005571266.2 | XP_005571323.1 | 1 - 2 | 460 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |