Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIPK1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005580401.2 | XP_005580458.1 | 2 - 3 | 1788 | 99 | |
XM_005580402.2 | XP_005580459.1 | 2 - 3 | 235 | 99 |