Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AZIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293755.1 | NP_001280684.1 | 6 - 7 | 959 | 95 | |
XM_015452248.1 | XP_015307734.1 | 6 - 7 | 932 | 95 | ||
XM_015452249.1 | XP_015307735.1 | 6 - 7 | 931 | 95 | ||
XM_015452251.1 | XP_015307737.1 | 7 - 8 | 998 | 95 | ||
XM_015452253.1 | XP_015307739.1 | 6 - 7 | 931 | 95 | ||
XM_015452254.1 | XP_015307740.1 | 5 - 6 | 754 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |