Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283398.1 | NP_001270327.1 | 8 - 9 | 1008 | 101 | |
XM_015432111.1 | XP_015287597.1 | 7 - 8 | 731 | 101 | ||
XM_015432112.1 | XP_015287598.1 | 6 - 7 | 660 | 101 | ||
XM_015432113.1 | XP_015287599.1 | 5 - 6 | 550 | 101 | ||
XM_015432114.1 | XP_015287600.1 | 9 - 10 | 846 | 101 | ||
XM_015432115.1 | XP_015287601.1 | 7 - 8 | 664 | 101 | ||
GenBank mRNA | AB070144.1 | - | 8 - 9 | 1009 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |