Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283872.1 | NP_001270801.1 | 2 - 3 | 233 | 97 | |
GenBank mRNA | AB070178.1 | - | 2 - 3 | 233 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |