Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acvr2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007396.4 | NP_031422.3 | 3 - 4 | 1037 | 93 | |
GenBank mRNA | AK041246.1 | - | 3 - 4 | 990 | 93 | |
AK143186.1 | - | 3 - 4 | 1112 | 93 | ||
BC138823.1 | - | 3 - 4 | 698 | 8861845 | 93 | |
M65287.1 | - | 3 - 4 | 444 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |