Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007762.4 | NP_031788.1 | 8 - 9 | 962 | 86 | |
GenBank mRNA | AB375515.1 | - | 8 - 9 | 772 | 86 | |
AF483484.1 | - | 8 - 9 | 772 | 86 | ||
AF483485.1 | - | 8 - 9 | 772 | 86 | ||
AK144430.1 | - | 8 - 9 | 1030 | 86 | ||
AK158806.1 | - | 8 - 9 | 963 | 86 | ||
BC103675.1 | - | 8 - 9 | 790 | 40046992 | 86 | |
BC104735.1 | - | 9 - 10 | 790 | 40046995 | 86 | |
BC105673.1 | - | 9 - 10 | 866 | 40046994 | 86 | |
BC105675.1 | - | 8 - 9 | 790 | 40046993 | 86 | |
FJ668671.1 | - | 7 - 8 | 652 | 86 | ||
X72305.1 | - | 8 - 9 | 803 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |