Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esrra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007953.2 | NP_031979.2 | 5 - 6 | 936 | 77 | |
GenBank mRNA | AK019222.1 | - | 5 - 6 | 897 | 77 | |
AK156371.1 | - | 5 - 6 | 912 | 77 | ||
BC034165.1 | - | 5 - 6 | 879 | 77 | ||
BC039774.1 | - | 5 - 6 | 948 | 77 | ||
BC138586.1 | - | 5 - 6 | 896 | 8861606 | 77 | |
BC171958.1 | - | 5 - 6 | 896 | 9054244 | 77 | |
U85259.1 | - | 5 - 6 | 903 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |