Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161373.1 | NP_001154845.1 | 4 - 5 | 1013 | 112 | |
NM_001161374.1 | NP_001154846.1 | 4 - 5 | 1013 | 112 | ||
NM_007977.2 | NP_032003.2 | 4 - 5 | 1013 | 112 | ||
GenBank mRNA | AJ627040.1 | - | 3 - 4 | 433 | 112 | |
AK051579.1 | - | 4 - 5 | 3289 | 112 | ||
BC138347.1 | - | 4 - 5 | 894 | 8861367 | 112 | |
BC145245.1 | - | 4 - 5 | 894 | 9053797 | 112 | |
L05573.1 | - | 4 - 5 | 1011 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |