Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122892.1 | NP_001116364.1 | 10 - 11 | 1557 | 62 | |
NM_001122893.1 | NP_001116365.1 | 11 - 12 | 1636 | 62 | ||
NM_008054.2 | NP_032080.2 | 11 - 12 | 1627 | 62 | ||
GenBank mRNA | AK156584.1 | - | 11 - 12 | 1638 | 62 | |
AK171646.1 | - | 11 - 12 | 1629 | 62 | ||
BC010554.1 | - | 10 - 11 | 1238 | 62 | ||
BC032149.1 | - | 10 - 11 | 1272 | 3602546 | 62 | |
BC092217.1 | - | 10 - 11 | 1553 | 6403711 | 62 | |
M27266.1 | - | 12 - 13 | 1339 | 62 | ||
U70324.1 | - | 11 - 12 | 1340 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |