Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008069.4 | NP_032095.1 | 7 - 8 | 833 | 88 | |
GenBank mRNA | BC130258.1 | - | 7 - 8 | 837 | 40142758 | 88 |
BC157920.1 | - | 7 - 8 | 899 | 9007350 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |