Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100007100.mm.1 |
NM_001109657.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007100.mm.1 |
TC1100000859.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007092.mm.1 |
NM_008088.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007092.mm.1 |
TC1100000859.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007093.mm.1 |
NM_008088.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007093.mm.1 |
TC1100000859.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007109.mm.1 |
NM_001109657.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007109.mm.1 |
NM_001277079.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007109.mm.1 |
NM_001277080.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007109.mm.1 |
NM_008088.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007109.mm.1 |
TC1100000859.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007102.mm.1 |
NM_001109657.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007102.mm.1 |
TC1100000859.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007105.mm.1 |
NM_001277079.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007105.mm.1 |
NM_001277080.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007105.mm.1 |
TC1100000859.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007107.mm.1 |
NM_001109657.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007107.mm.1 |
NM_001277079.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007107.mm.1 |
NM_001277080.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007107.mm.1 |
NM_008088.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007107.mm.1 |
TC1100000859.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007101.mm.1 |
NM_001109657.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007101.mm.1 |
TC1100000859.mm.1 |
MTA TC |