Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008156.2 | NP_032182.2 | 2 - 3 | 412 | 94 | |
GenBank mRNA | AF050666.1 | - | 2 - 3 | 366 | 94 | |
AK004957.2 | - | 1 - 2 | 116 | 94 | ||
AY081194.1 | - | 1 - 2 | 139 | 94 | ||
AY333960.1 | - | 1 - 2 | 96 | 94 | ||
BC019146.1 | - | 1 - 2 | 125 | 5052822 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |