Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008192.3 | NP_032218.2 | 12 - 13 | 2513 | 74 | |
GenBank mRNA | BI738500.1 | - | 3 - 4 | 465 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |