Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277290.1 | NP_001264219.1 | 3 - 4 | 727 | 54 | |
NM_008238.2 | NP_032264.1 | 3 - 4 | 730 | 54 | ||
GenBank mRNA | BC108980.1 | - | 2 - 3 | 615 | 40047616 | 54 |
BC108981.1 | - | 2 - 3 | 618 | 40047617 | 54 | |
X81593.1 | - | 3 - 4 | 680 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |