Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110252.2 | NP_001103722.1 | 12 - 13 | 1354 | 70 | |
NM_001276269.1 | NP_001263198.1 | 11 - 12 | 1231 | 70 | ||
NM_008281.4 | NP_032307.2 | 11 - 12 | 1294 | 70 | ||
GenBank mRNA | AF030065.1 | - | 11 - 12 | 1253 | 70 | |
AK002694.1 | - | 12 - 13 | 1309 | 70 | ||
AK156553.1 | - | 11 - 12 | 1233 | 70 | ||
AY234104.1 | - | 10 - 11 | 1359 | 70 | ||
BC138809.1 | - | 12 - 13 | 1198 | 8861831 | 70 | |
BC145413.1 | - | 12 - 13 | 1198 | 9053971 | 70 | |
CB949755.1 | - | 4 - 5 | 465 | 70 | ||
CJ237993.1 | - | 2 - 3 | 171 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |