Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il10ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008348.2 | NP_032374.1 | 3 - 4 | 449 | 62 | |
GenBank mRNA | AK137663.1 | - | 3 - 4 | 450 | 62 | |
AK149680.1 | - | 3 - 4 | 444 | 62 | ||
AK149899.1 | - | 2 - 3 | 332 | 62 | ||
AK150022.1 | - | 3 - 4 | 446 | 62 | ||
AK150524.1 | - | 3 - 4 | 453 | 62 | ||
AK153120.1 | - | 3 - 4 | 453 | 62 | ||
AK159566.1 | - | 3 - 4 | 446 | 62 | ||
L12120.1 | - | 3 - 4 | 454 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |