Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il2ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008367.3 | NP_032393.3 | 5 - 6 | 855 | 58 | |
GenBank mRNA | AK037369.1 | - | 5 - 6 | 841 | 58 | |
AK088594.1 | - | 5 - 6 | 797 | 58 | ||
BC114437.1 | - | 5 - 6 | 761 | 40054387 | 58 | |
K02891.1 | - | 5 - 6 | 738 | 58 | ||
M54934.1 | - | 5 - 6 | 658 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |