Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048177.2 | NP_001041642.1 | 17 - 18 | 2773 | 91 | |
NM_008413.3 | NP_032439.2 | 18 - 19 | 2856 | 91 | ||
GenBank mRNA | BC054807.1 | - | 17 - 18 | 2536 | 6838318 | 91 |
BC059834.1 | - | 17 - 18 | 2536 | 6838318 | 91 | |
L16956.1 | - | 17 - 18 | 2525 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |