Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008522.3 | NP_032548.2 | 5 - 6 | 676 | 66 | |
GenBank mRNA | AK036491.1 | - | 5 - 6 | 677 | 66 | |
AK144556.1 | - | 5 - 6 | 677 | 66 | ||
AK150799.1 | - | 5 - 6 | 677 | 66 | ||
AK151822.1 | - | 5 - 6 | 677 | 66 | ||
AK164781.1 | - | 5 - 6 | 678 | 66 | ||
BC006904.1 | - | 5 - 6 | 684 | 3600033 | 66 | |
BC008530.1 | - | 5 - 6 | 657 | 66 | ||
CT010339.1 | - | 5 - 6 | 642 | 66 | ||
D88510.1 | - | 5 - 6 | 676 | 66 | ||
FJ538998.1 | - | 5 - 6 | 642 | 66 | ||
J03298.1 | - | 5 - 6 | 645 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |