Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mug1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008645.3 | NP_032671.2 | 17 - 18 | 2164 | 141 | |
GenBank mRNA | AK144255.1 | - | 17 - 18 | 2128 | 141 | |
AK144541.1 | - | 17 - 18 | 2122 | 141 | ||
AK149376.1 | - | 17 - 18 | 2128 | 141 | ||
BC036299.1 | - | 17 - 18 | 2110 | 141 | ||
BC049868.1 | - | 17 - 18 | 2162 | 141 | ||
BC051037.1 | - | 17 - 18 | 2164 | 4195193 | 141 | |
M65736.1 | - | 17 - 18 | 2105 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105413 | BC036299.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105413 | BC049868.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105413 | BC051037.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105413 | M65736.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105413 | NM_008645.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724682 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 42426 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 527622 | AK009960.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 527622 | AK031533.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 527622 | AK081728.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 527622 | BC037466.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 527622 | BC043033.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 527622 | BC043081.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 527622 | BC048787.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 527622 | NM_027171.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 44429 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931132 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 169511 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 166273 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12924 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134064 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174825 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105413 | BC036299.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105413 | BC049868.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105413 | BC051037.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105413 | M65736.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105413 | NM_008645.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P182631 | BC036299.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P182631 | BC049868.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P182631 | BC051037.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P182631 | M65736.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P182631 | NM_008645.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24806184 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24806195 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24806176 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24806158 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24806162 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06051091 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174825 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06051078 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06051068 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P134064 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01435253 | - | - |
Whole Mouse Genome G4122A | A_51_P182631 | BC036299.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P182631 | BC049868.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P182631 | BC051037.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P182631 | M65736.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P182631 | NM_008645.3 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105926 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114554 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105765 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174825 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06051078 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P134064 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05470110 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05470111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806338 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806409 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806500 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806486 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806176 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806484 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24806244 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24806178 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24806210 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05864209 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05193302 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05470110 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05699253 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24806145 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24806148 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24806195 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05699252 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05864212 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05470111 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24806162 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05200545 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24806207 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05470099 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24806190 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10541425 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541470 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541421 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541468 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541451 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541465 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541422 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541418 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541453 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541454 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541464 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541414 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541415 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541452 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541456 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541459 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541467 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541416 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541472 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541411 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541466 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541413 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541473 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541476 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541478 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541420 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541419 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541424 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541417 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541462 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541412 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10541481 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1417835_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10541448 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10541410 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10541480 | - | - |
Mouse Genome 430 2.0 Array | 1448854_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1417835_at | - | - |
Mouse Genome 430 2.0 Array | 1417835_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1448854_s_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17836_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17836_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600011000.mm.1 | TC0600001373.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600011000.mm.1 | NM_008645.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011000.mm.1 | NM_008645.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011014.mm.1 | TC0600001373.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600011014.mm.1 | NM_008645.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011014.mm.1 | NM_008645.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600010998.mm.1 | TC0600001373.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600010998.mm.1 | NM_008645.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600010998.mm.1 | NM_008645.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600010999.mm.1 | TC0600001373.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600010999.mm.1 | NM_008645.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600010999.mm.1 | NM_008645.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011010.mm.1 | TC0600001373.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600011010.mm.1 | NM_008645.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011010.mm.1 | NM_008645.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011011.mm.1 | TC0600001373.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600011011.mm.1 | NM_008645.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011011.mm.1 | NM_008645.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011013.mm.1 | TC0600001373.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600011013.mm.1 | NM_008645.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011013.mm.1 | NM_008645.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011018.mm.1 | TC0600001373.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600011018.mm.1 | NM_008645.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011018.mm.1 | NM_008645.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011003.mm.1 | TC0600001373.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600011003.mm.1 | NM_008645.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011003.mm.1 | NM_008645.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011004.mm.1 | TC0600001373.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600011004.mm.1 | NM_008645.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011004.mm.1 | NM_008645.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011006.mm.1 | TC0600001373.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600011006.mm.1 | NM_008645.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011006.mm.1 | NM_008645.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011008.mm.1 | TC0600001373.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600011008.mm.1 | NM_008645.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011008.mm.1 | NM_008645.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011002.mm.1 | TC0600001373.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600011002.mm.1 | NM_008645.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011002.mm.1 | NM_008645.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600010996.mm.1 | TC0600001373.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600010996.mm.1 | NM_008645.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600010996.mm.1 | NM_008645.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600010997.mm.1 | TC0600001373.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600010997.mm.1 | NM_008645.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600010997.mm.1 | NM_008645.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011005.mm.1 | TC0600001373.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600011005.mm.1 | NM_008645.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011005.mm.1 | NM_008645.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011015.mm.1 | TC0600001373.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600011015.mm.1 | NM_008645.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011015.mm.1 | NM_008645.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011007.mm.1 | TC0600001373.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600011007.mm.1 | NM_008645.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011007.mm.1 | NM_008645.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011009.mm.1 | TC0600001373.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600011009.mm.1 | NM_008645.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011009.mm.1 | NM_008645.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011012.mm.1 | TC0600001373.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600011012.mm.1 | NM_008645.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011012.mm.1 | NM_008645.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011016.mm.1 | TC0600001373.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600011016.mm.1 | NM_008645.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600011016.mm.1 | NM_008645.3 | Refseq |
Mouse Expression Set 430 | 1417835_PM_at | - | - |
Mouse Expression Set 430 | 1448854_PM_s_at | - | - |
Mouse Expression Set 430 | 1417835_at | - | - |
Mouse Expression Set 430 | 1448854_s_at | - | - |
Murine Genome U74v2 Set | 92837_PM_f_at | - | - |
Murine Genome U74v2 Set | 92837_f_at | - | - |
Murine Genome U74v2 Set | 97680_PM_at | - | - |
Murine Genome U74v2 Set | 97680_at | - | - |
Murine Genome U74v2 Set | 135778_at | - | - |
Murine Genome U74v2 Set | 135778_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1448854_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1417835_at | - | - |
Mouse Genome 430A 2.0 Array | 1417835_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1448854_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417835_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417835_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 314374 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 610706 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3190273 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 610706 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3170497 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000610706 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003190273 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003170497 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2683432 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1224954 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1224954 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2683432 | - | - |
mouse-6 v1.1 (Illumina) | scl017836.18_18-S | - | - |
mouse-6 v1.1 (Illumina) | scl0017836.1_208-S | - | - |
mouse-6 v1.1 (Illumina) | scl017836.34_27-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3170497 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 610706 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3190273 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0017836.1_208-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl017836.18_18-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl017836.34_27-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 610706 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3190273 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3170497 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2774563 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2774563 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1224954 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2683432 | - | - |