Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008737.2 | NP_032763.2 | 8 - 9 | 1770 | 104 | |
GenBank mRNA | AK082992.1 | - | 8 - 9 | 1823 | 104 | |
AK141964.1 | - | 8 - 9 | 1562 | 104 | ||
AK148372.1 | - | 8 - 9 | 1746 | 104 | ||
AK149752.1 | - | 3 - 4 | 351 | 104 | ||
BC060129.1 | - | 8 - 9 | 1550 | 6409596 | 104 | |
D50086.1 | - | 8 - 9 | 1630 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |