Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161825.1 | NP_001155297.1 | 5 - 6 | 644 | 76 | |
NM_008822.2 | NP_032848.1 | 6 - 7 | 722 | 76 | ||
GenBank mRNA | AK004687.1 | - | 6 - 7 | 707 | 76 | |
AK160195.1 | - | 6 - 7 | 686 | 76 | ||
BC132213.1 | - | 6 - 7 | 695 | 40130490 | 76 | |
BC132503.1 | - | 6 - 7 | 695 | 40130780 | 76 | |
BC145300.1 | - | 5 - 6 | 617 | 9053854 | 76 | |
U69171.1 | - | 6 - 7 | 713 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |