Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008856.3 | NP_032882.2 | 9 - 10 | 1393 | 71 | |
GenBank mRNA | AK076352.1 | - | 9 - 10 | 1592 | 71 | |
AK145817.1 | - | 9 - 10 | 1639 | 71 | ||
AK155951.1 | - | 10 - 11 | 1736 | 71 | ||
AK164044.1 | - | 9 - 10 | 1592 | 71 | ||
BC031121.1 | - | 9 - 10 | 1393 | 5059633 | 71 | |
D90242.1 | - | 9 - 10 | 1330 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |