Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008878.2 | NP_032904.1 | 7 - 8 | 765 | 65 | |
GenBank mRNA | AK077563.1 | - | 7 - 8 | 767 | 65 | |
AK090208.1 | - | 7 - 8 | 749 | 65 | ||
BC026756.1 | - | 7 - 8 | 739 | 5124817 | 65 | |
GQ896353.1 | - | 6 - 7 | 716 | 65 | ||
GQ896354.1 | - | 6 - 7 | 716 | 65 | ||
Z36774.1 | - | 7 - 8 | 752 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |