Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253892.1 | NP_001240821.1 | 4 - 5 | 647 | 59 | |
NM_009207.3 | NP_033233.2 | 5 - 6 | 843 | 59 | ||
GenBank mRNA | AK145930.1 | - | 5 - 6 | 768 | 59 | |
AK147858.1 | - | 5 - 6 | 799 | 59 | ||
AK180093.1 | - | 1 - 2 | 124 | 59 | ||
AK219021.1 | - | 1 - 2 | 84 | 59 | ||
BC054102.1 | - | 4 - 5 | 652 | 30017310 | 59 | |
J04036.1 | - | 5 - 6 | 758 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |