Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tle3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083927.1 | NP_001077396.1 | 2 - 3 | 1193 | 69 | |
NM_001083928.1 | NP_001077397.1 | 2 - 3 | 1193 | 69 | ||
NM_009389.2 | NP_033415.2 | 2 - 3 | 1193 | 69 | ||
GenBank mRNA | AK122524.1 | - | 2 - 3 | 701 | 69 | |
AK158785.1 | - | 1 - 2 | 516 | 69 | ||
BC056465.1 | - | 2 - 3 | 174 | 5709252 | 69 | |
X73360.1 | - | 1 - 2 | 376 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |