Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tle3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083927.1 | NP_001077396.1 | 11 - 12 | 2016 | 62 | |
NM_001083928.1 | NP_001077397.1 | 11 - 12 | 1986 | 62 | ||
NM_009389.2 | NP_033415.2 | 11 - 12 | 1986 | 62 | ||
GenBank mRNA | AK031322.1 | - | 9 - 10 | 793 | 62 | |
AK122524.1 | - | 11 - 12 | 1494 | 62 | ||
AK147635.1 | - | 3 - 4 | 5213 | 62 | ||
AK158785.1 | - | 10 - 11 | 1306 | 62 | ||
BC056465.1 | - | 11 - 12 | 997 | 5709252 | 62 | |
X73360.1 | - | 10 - 11 | 1169 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |