Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vdr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009504.4 | NP_033530.2 | 4 - 5 | 423 | 58 | |
GenBank mRNA | AK030339.1 | - | 4 - 5 | 424 | 58 | |
AK154647.1 | - | 4 - 5 | 398 | 58 | ||
AK156789.1 | - | 4 - 5 | 439 | 58 | ||
BC006716.1 | - | 4 - 5 | 389 | 3710866 | 58 | |
CT010294.1 | - | 2 - 3 | 283 | 58 | ||
D31969.1 | - | 4 - 5 | 391 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Nuclear Receptor Mammalian Gene Collection Mus musculus Probe spans exons |