Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200009358.mm.1 |
NM_009511.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009358.mm.1 |
NM_009511.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009358.mm.1 |
TC1200001299.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009353.mm.1 |
NM_009511.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009353.mm.1 |
NM_009511.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009353.mm.1 |
TC1200001299.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009354.mm.1 |
NM_009511.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009354.mm.1 |
NM_009511.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009354.mm.1 |
TC1200001299.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009357.mm.1 |
NM_009511.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009357.mm.1 |
NM_009511.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009357.mm.1 |
TC1200001299.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009355.mm.1 |
NM_009511.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009355.mm.1 |
NM_009511.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009355.mm.1 |
TC1200001299.mm.1 |
MTA TC |